Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113738 Supplier Invitrogen™ Supplier No. PA5113738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGHQEAPERE RRSEEPPISL DLTFHLLREV LEMARAEQLA QQAHSNRKLM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 92%; Zebrafish: 93%.

Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRF
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CRH
Gene Accession No. P06850
Gene Alias Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF
Gene Symbols CRH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human CRH (aa 142-191).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1392
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.