Learn More
Invitrogen™ CRF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116529
Description
Positive Samples: SH-SY5Y, Mouse liver, Mouse brain, Rat liver, Rat brain Immunogen sequence: LLSRGPVPGA RQAPQHPQPL DFFQPPPQSE QPQQPQARPV LLRMGEEYFL RLGNLNKSPA APLSPASSLL AGGSGSRPSP EQATANFFRV LLQQLLLPRR SLDSPAALAE RGARNALGGH QEAPERERRS EEPPISLDLT FHLLREVLEM ARAEQLAQQA HSNRKLMEII.
Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.
Specifications
CRF | |
Polyclonal | |
Unconjugated | |
CRH | |
Corticoliberin; corticotropin releasing factor; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; corticotropin-releasing factor; Corticotropin-releasing factor receptor 2; corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF2; CRF2 receptor, beta isoform; CRF2R; CRFR2; CRF-R-2; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF | |
Rabbit | |
Affinity chromatography | |
RUO | |
12918, 1392, 81648 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01143, P06850, Q8CIT0 | |
CRH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-194 of human CRH (NP_0007471). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.