Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980174
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0174 Supplier Bioss Supplier No. BS0246R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Corticotropin Releasing Factor (CRF), a 41 amino acids neuropeptide, belongs to a family of structurally related peptides including urocortin, sauvagine and urotensin I. CRF is critical in the regulation of the pituitary-adrenal gland axis, and in endocrine, autonomic and behavioral responses to stress. CRF is widely distributed in the brain and peripheral nervous system with the highest levels in the hypothalamic paraventricular nucleus.

Specifications

Antigen CRF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Crhr2
Gene Accession No. P06850, Q13324
Gene Alias Corticoliberin; corticotrophin releasing hormone receptor 2; corticotropin releasing factor; corticotropin releasing factor receptor type 2 beta; corticotropin releasing hormone; corticotropin releasing hormone receptor 2; Corticotropin-releasing factor; corticotropin-releasing factor 2; Corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; Corticotropin-releasing hormone; Corticotropin-releasing hormone receptor 2; CRF; CRF 2 receptor; CRF2; CRF2 receptor, beta isoform; CRF2R; CRF-R 2; CRFR2; CRF-R2; CRF-R-2; CRFR-2; CRFR2alpha; CRFR2beta; CRF-RB; CRH; CRH receptor 2 variant B; CRH1; CRH2R; CRH-R 2; CRHR2; CRH-R2; CRH-R-2; Gm1347; HM-CRF
Gene Symbols CRH, Crhr2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1392, 1395
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.