Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRG-L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519534
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519534 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519534 Supplier Invitrogen™ Supplier No. PA519534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with human based on sequence homology.

CRG-L2 is a protein that contains olfactomedin-like and collagen-like domains. The encoded protein, which exists in both transmembrane and secreted forms, promotes formation of the nodes of Ranvier in the peripheral nervous system. Mutations in this gene cause a form of lethal congenital contracture syndrome in human patients. Autoantibodies to the encoded protein have been identified in sera form patients with multifocal motor neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRG-L2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene Gldn
Gene Accession No. Q80WL1, Q8BMF8
Gene Alias cancer related gene - liver 2; cancer-related gene liver 2 protein; Clom; collomin; COLM; colmedin; Crgl2; CRG-L2; Crlg2; FLJ23917; Gldn; Gliomedin; Gliomedin shedded ectodomain; olfactomedin-related protein; UNC-112; UNQ9339/PRO34011
Gene Symbols Gldn
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 250 - 350 of Rat Gliomedin.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235379, 315675
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.