Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRHBP Monoclonal Antibody (D2), Invitrogen™

Catalog No. FEMA550972
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550972 20 μL
FEMA550971 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550972 Supplier Invitrogen™ Supplier No. MA550972
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Response to stress in mammals requires an intact hypothalamic-pituitary-adrenal axis. The proximal part of the response is mediated by secretion of corticotropin-releasing hormone (CRH) by the paraventricular nucleus of the hypothalamus. CRH is a 41 amino acid peptide derived by enzymatic cleavage from a 191 amino acid preprohormone. CRH is produced not only in the hypothalamus but also in peripheral tissues, such as T lymphocytes; it is highly expressed in human placenta. Glucocorticoids stimulate placental CRH synthesis and secretion in primary cultures of human placenta. This stimulation is in contrast to the glucocorticoid suppression of CRH expression in hypothalamus. The gene which encodes CRH maps to human chromosome 8q13. Human plasma contains a CRH-binding protein, CRH-BP (also designated CRF-BP) which inactivates CRH and which may prevent inappropriate pituitary-adrenal stimulation in pregnancy. The gene which encodes CRF-BP maps to human chromosome 5q11.2-q13.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRHBP
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CRHBP
Gene Accession No. P24388
Gene Alias corticotropin releasing factor binding protein; corticotropin releasing hormone binding protein; corticotropin releasing hormone-binding protein; corticotropin-releasing factor-binding protein; Corticotropin-releasing hormone-binding protein; CRF-binding protein; CRFBP; CRF-BP; Crhbp; CRH-BP
Gene Symbols CRHBP
Host Species Mouse
Immunogen Recombinant Rat CRHBP protein, Tyr25-Leu322 (Accession #P24388).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29625
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.