Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRHR1/CRF1 Antibody (343919), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3930PECY55
Description
CRHR1/CRF1 Monoclonal antibody specifically detects CRHR1/CRF1 in Human samples. It is validated for Flow CytometrySpecifications
CRHR1/CRF1 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
343919 | |
Flow Cytometry | |
corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor | |
NS0 mouse myeloma cell line transfected with human CRHR-1, Ser24-Val415, Accession # NP_004373 | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
1394 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction