Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533447
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533447 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533447 Supplier Invitrogen™ Supplier No. PA533447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is not recommended for use in ICC applications. Percent identity with other species by BLAST analysis: Human, Monkey (100%) Marmoset (94%) Rat (82%).

Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRHR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CRHR1
Gene Accession No. P34998, P35353
Gene Alias corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; corticotropin releasing hormone receptor 1; corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; corticotropin-releasing hormone receptor 1; CRF 1 receptor; Crf receptor 1; Crf type 1; CRF1; Crf1 receptor; CRFR; CRF-R; CRFR1; CRF-R1; CRF-R-1; CRFR-1; CRF-R1alpha; CRH receptor 1; CRH receptor subtype-1 isoform-3; Crhr; CRH-R 1; CRHR1; CRH-R1; CRH-R-1; CRHR1f; CRH-R1h; CRHR1L; seven transmembrane helix receptor
Gene Symbols CRHR1
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from N-terminal extracellular domain of human CRF1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1394, 58959
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.