Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRHR1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989742
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9742 Supplier Bioss Supplier No. BS10248R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.

Specifications

Antigen CRHR1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene CRHR1
Gene Accession No. P34998
Gene Alias corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; corticotropin releasing hormone receptor 1; Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF 1 receptor; Crf receptor 1; Crf type 1; CRF1; Crf1 receptor; CRFR; CRF-R; CRFR1; CRF-R1; CRF-R-1; CRFR-1; CRF-R1alpha; CRH receptor 1; CRH receptor subtype-1 isoform-3; CRHR; CRH-R 1; CRHR1; CRH-R1; CRH-R-1; CRHR1f; CRH-R1h; CRHR1L; seven transmembrane helix receptor
Gene Symbols CRHR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRHR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1394
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.