Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRHR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 720291
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-029-1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-029-1 Supplier Invitrogen™ Supplier No. 720291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Pig, Rat and Mouse.

Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRHR2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Gene CRHR2
Gene Accession No. Q13324, Q60748
Gene Alias corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin releasing hormone receptor 2; corticotropin-releasing factor 2; corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; corticotropin-releasing hormone receptor 2; corticotropin-releasing hormone receptor type 2 variant 2; CRF 2 receptor; CRF2; CRF2 receptor, beta isoform; Crf2r; CRF-R 2; CRFR2; CRF-R2; CRF-R-2; CRFR-2; CRFR2alpha; CRFR2beta; CRF-RB; CRH receptor 2 variant B; Crh2 receptor; CRH2R; CRH-R 2; Crhr2; CRH-R2; CRH-R-2; HM-CRF; putative corticotropin-releasing hormone receptor type 2
Gene Symbols CRHR2
Host Species Rabbit
Immunogen Peptide corresponding to Human CRHR2 (aa 24-38).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12922, 1395
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.