Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRHR2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528755 Supplier Thermo Scientific Supplier No. PA528755

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116. Predicted reactivity: Pig (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRHR2
Applications Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CRHR2
Gene Accession No. Q13324
Gene Alias corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin releasing hormone receptor 2; corticotropin-releasing factor 2; corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; corticotropin-releasing hormone receptor 2; corticotropin-releasing hormone receptor type 2 variant 2; CRF 2 receptor; CRF2; CRF2 receptor, beta isoform; Crf2r; CRF-R 2; CRFR2; CRF-R2; CRF-R-2; CRFR-2; CRFR2alpha; CRFR2beta; CRF-RB; CRH receptor 2 variant B; Crh2 receptor; CRH2R; CRH-R 2; Crhr2; CRH-R2; CRH-R-2; HM-CRF; putative corticotropin-releasing hormone receptor type 2
Gene Symbols CRHR2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human CRF-RII
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1395
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.