Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRHR2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984972
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4972 Supplier Bioss Supplier No. BS2792R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.

Specifications

Antigen CRHR2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CRHR2
Gene Accession No. Q13324
Gene Alias BOS_4309; corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin releasing hormone receptor 2; corticotropin-releasing factor 2; Corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; Corticotropin-releasing hormone receptor 2; CRF 2 receptor; CRF2; CRF2 receptor, beta isoform; CRF2R; CRF-R 2; Crfr2; CRF-R2; CRF-R-2; CRFR-2; CRFR2alpha; CRFR2beta; CRF-RB; CRH receptor 2 variant B; Crh2 receptor; CRH2R; CRH-R 2; Crhr2; CRH-R2; CRH-R-2; HM-CRF; putative corticotropin-releasing hormone receptor type 2
Gene Symbols CRHR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRHR2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1395
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.