Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRHSP-24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580407
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CARHSP1.
CRHSP-24 is a protein coding gene.
Specifications
| CRHSP-24 | |
| Polyclonal | |
| Unconjugated | |
| CARHSP1 | |
| 1200011K09Rik; calcineurin substrate CRHSP-24; calcium regulated heat stable protein 1; calcium regulated heat stable protein 1, 24kDa; calcium-regulated heat stable protein 1; calcium-regulated heat-stable protein (24kD); calcium-regulated heat-stable protein 1; Calcium-regulated heat-stable protein of 24 kDa; Carhsp1; CRHSP24; Crhsp-24; CSDC1; D16Ertd465e | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y2V2 | |
| CARHSP1 | |
| Recombinant Human CARHSP1 protein (Met1-Ser147). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction