Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRHSP-24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521471
Description
Recommended positive controls: HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (96%), Rhesus Monkey (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CRHSP-24 is a protein coding gene.
Specifications
| CRHSP-24 | |
| Polyclonal | |
| Unconjugated | |
| CARHSP1 | |
| 1200011K09Rik; calcineurin substrate CRHSP-24; calcium regulated heat stable protein 1; calcium regulated heat stable protein 1, 24kDa; calcium-regulated heat stable protein 1; calcium-regulated heat-stable protein (24kD); calcium-regulated heat-stable protein 1; Calcium-regulated heat-stable protein of 24 kDa; Carhsp1; CRHSP24; Crhsp-24; CSDC1; D16Ertd465e | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23589, 52502 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.71 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9CR86, Q9Y2V2 | |
| CARHSP1 | |
| Recombinant protein encompassing a sequence within the center region of human CRHSP-24. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction