Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CRID3 sodium salt
GREENER_CHOICESDP

Catalog No. 547950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-795-0 50 mg
54-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-795-0 Supplier R&D Systems™ Supplier No. 5479/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent NLRP3 inflammasome inhibitor; inhibits IL-1β production

CRID3 sodium salt is a potent NLRP3 inflammasome inhibitor; closes the active conformation of NLRP3 to the inactive state. Directly interacts with the Walker B motif within the NLRP3 NACHT domain. Inhibits IL-1β, IL-18 and IL-1α production (IC50 values are 7.2, 10.3 and 12-18 nM, respectively). Selective for NLRP3 over NLRC4 inflammasome and Toll-like receptor signaling. Reduces severity of experimental autoimmune encephalomyelitis, skin inflammation and airway inflammation in mice. Also glutathione S-transferase omega 1 inhibitor. Orally bioavailable.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type CRID3 sodium salt
Purity 98%
Molecular Weight (g/mol) 426.46

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.