Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRIF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CRIF1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CRIF1 Polyclonal specifically detects CRIF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CRIF1 | |
Polyclonal | |
Rabbit | |
Cancer | |
CKBBP2, CKII beta-associating protein, CR6 interacting factor 1, CRIF1p53-responsive gene 6 protein, growth arrest and DNA damage-inducible proteins-interacting protein 1, growth arrest and DNA-damage-inducible, gamma interacting protein 1, MGC4667, MGC4758, papillomavirus L2 interacting nuclear protein 1, Papillomavirus L2-interacting nuclear protein 1, Plinp1, PLINP-1CKII beta binding protein 2, PRG6CR6-interacting factor 1 | |
GADD45GIP1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
90480 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ELEAEEREWYPSLATMQESLRVKQLAEEQKRRER | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title