Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIg Mouse, anti-Mouse, R718, Clone: 17C9, BD OptiBuild™

Catalog No. BDB752146 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752146 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752146 Supplier BD Biosciences Supplier No. 752146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 17C9 monoclonal antibody specifically recognizes Complement receptor of the immunoglobulin superfamily (CRIg) which is encoded by the Vsig4 (V-set and immunoglobulin domain containing 4) gene. CRIg is a ∼45 kDa single-pass type I transmembrane glycoprotein that contains a variable (V-type) immunoglobulin domain in its extracellular region which is required for binding complement components and promoting phagocytosis. CRIg is expressed on Kupffer cells and tissue resident macrophages, including a subset of resident peritoneal macrophages. CRIg binds complement fragments C3b and iC3b and plays important roles in innate and adaptive immunity. This complement receptor is involved in regulation of the complement cascade and serves as a phagocytic receptor in the clearance of pathogens, immune complexes, and apoptotic cells. CRIg-positive macrophages can reportedly function in the negative regulation of T-cell proliferation and IL-2 production.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIg
Applications Flow Cytometry
Classification Monoclonal
Clone 17C9
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.