Learn More
CRIM1 Antibody (MM0202-16G1), Alexa Fluor™ 532, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212207AF532
Catalog No. NP212207A32
Only null left
Add to Cart
Description
Description
CRIM1 Monoclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CRIM1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CRIM1
Human recombinant CRIM1 (extracellular domain)
Primary
Human
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0202-16G1
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52
Mouse
0.1 mL
51232
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.