Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534410
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534410 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534410 Supplier Thermo Scientific Supplier No. PA534410
Add to Cart
Add to Cart

CRIM1 Polyclonal Antibody

CRIM1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CRIM1
Gene Accession No. Q9NZV1
Gene Alias AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330
Gene Symbols CRIM1
Host Species Rabbit
Immunogen A 17 amino acid synthetic peptide near the carboxy terminus of human Crim1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51232
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.