Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534411
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534411 Supplier Invitrogen™ Supplier No. PA534411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-34411 can be used with blocking peptide PEP-1453.

CRIM2, also known as Kielin/chordin-like protein (KCP), is a novel enhancer of BMP signaling. It binds to BMP7, a morphogen that is important for kidney development and part of the kidney's physiological response to repair of acute kidney injury, enhancing its binding to its receptor. Because of this, CRIM2 has been suggested as a potential antifibrotic therapy in acute and chronic kidney injury. CRIM2 can also interact directly with either activin or TGF-Beta1, suppressing activin- and TGF-Beta1-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KCP
Gene Accession No. Q3U492, Q6ZWJ8
Gene Alias AW060220; CRIM2; CRIM-2; cysteine rich BMP regulator 2 (chordin like); cysteine rich BMP regulator 2 (chordin-like); cysteine-rich BMP regulator 2; Cysteine-rich motor neuron 2 protein; Gm793; KCP; Kcp1; KCP-1; kielin; kielin/chordin-like protein; Kielin/chordin-like protein 1; NET67; RGD1561119
Gene Symbols KCP
Host Species Rabbit
Immunogen A 15 amino acid synthetic peptide near the carboxy terminus of human CRIM2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296952, 333088, 375616
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.