Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Crimp seals with PTFE/Rubber Septa Gold Aluminum Seal

Catalog No. p-7204482 Shop All MilliporeSigma Supelco Products
Click to view available options
Material:
Aluminum (Seal), TFE, Butyl Rubber

Closure type crimp seal

  • The PTFE/rubber septa are compatible with most organic solvents until pierced and are not recommended for aromatics, carbon disulfides, chlorinated solvents, hydrocarbons.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 11 mm
Color Red, Gold
Material Aluminum (Seal), TFE, Butyl Rubber
Septum Rubber
No. per Pack 100
Thickness (Metric) 1.0 mm
For Use With (Equipment) 11 mm Crimp Top Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.