Learn More
Description
- 1 in. face width
- Max safe speed: 6,000 RPM
Specifications
Specifications
| Product Type | Wheel Brush |
| Diameter (English) | 6 in. |
| For Use With (Equipment) | 1/2 in. Rotating Shaft |
| Quantity | 1 Ea. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
