Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559984 Supplier Invitrogen™ Supplier No. PA559984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCYAAMFGPK GFGRGGAESH TFK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

CRIP1 is a cysteine-rich intestinal protein (CRIP) that belongs to the LIM/double zinc finger protein family, members of which include cysteine- and glycine-rich protein-1 (CSRP1 MIM 123876), rhombotin-1 (RBTN1 MIM 186921), rhombotin-2 (RBTN2 MIM 180385), and rhombotin-3 (RBTN3 MIM 180386). CRIP may be involved in intestinal zinc transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRIP1
Gene Accession No. P50238
Gene Alias CRHP; Crip; CRIP1; CRP1; CRP-1; cysteine rich intestinal protein; cysteine rich protein 1; cysteine-rich heart protein; Cysteine-rich intestinal protein; cysteine-rich protein 1; cysteine-rich protein 1 (intestinal)
Gene Symbols CRIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRIP1. Recombinant protein control fragment (Product #RP-99286).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1396
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.