Learn More
Invitrogen™ CRIP2 Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | CRIP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.55 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Crip2 |
| Gene Accession No. | P52943 |
| Gene Alias | 0610010I23Rik; AW743261; C77570; CRIP; Crip2; Crp; CRP2; CRP-2; CRP4; cysteine rich protein 2; Cysteine-rich intestinal protein; cysteine-rich protein 2; ESP1; heart LIM protein; Hlp; LIM domain protein ESP1/CRP2; LIM only protein HLP; LIM/double zinc-finger protein; Protein ESP1 |
| Gene Symbols | Crip2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.