Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583622 Supplier Invitrogen™ Supplier No. PA583622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAGSYIYEKP LAEGPQVTGP IEVPAARAEE RKASGPPKGP SRASSVTTFT GEPNTCPRCS KKVYFAE Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.

CRIP2 is a 208-amino acid protein containing 2 LIM domains and shares 93% amino acid sequence identity with its rat homolog, called Esp1 or Crp2. Northern blot analysis showed widespread tissue expression of the 1.3-kb CRIP2 mRNA, with the highest level of expression found in heart. In testis, a second 1.7-kb mRNA was also detected.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Crip2
Gene Accession No. P52943
Gene Alias 0610010I23Rik; AW743261; C77570; CRIP; Crip2; Crp; CRP2; CRP-2; CRP4; cysteine rich protein 2; Cysteine-rich intestinal protein; cysteine-rich protein 2; ESP1; heart LIM protein; Hlp; LIM domain protein ESP1/CRP2; LIM only protein HLP; LIM/double zinc-finger protein; Protein ESP1
Gene Symbols Crip2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRIP2. Recombinant protein control fragment (Product #RP-99950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1397
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.