Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597619 Supplier Invitrogen™ Supplier No. PA597619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRIP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CRIP2 is a 208-amino acid protein containing 2 LIM domains and shares 93% amino acid sequence identity with its rat homolog, called Esp1 or Crp2. Northern blot analysis showed widespread tissue expression of the 1.3-kb CRIP2 mRNA, with the highest level of expression found in heart. In testis, a second 1.7-kb mRNA was also detected.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Crip2
Gene Accession No. P36201, P52943, Q9DCT8
Gene Alias 0610010I23Rik; AW743261; C77570; CRIP; Crip2; Crp; CRP2; CRP-2; CRP4; cysteine rich protein 2; Cysteine-rich intestinal protein; cysteine-rich protein 2; ESP1; heart LIM protein; Hlp; LIM domain protein ESP1/CRP2; LIM only protein HLP; LIM/double zinc-finger protein; Protein ESP1
Gene Symbols Crip2
Host Species Rabbit
Immunogen Recombinant Human Cysteine-rich protein 2 protein (1-208AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1397, 338401, 68337
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.