Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRIPT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566039
Description
Immunogen sequence: CAYKKGICAMC GKKVLDTKNY KQTSV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.
Specifications
CRIPT | |
Polyclonal | |
Unconjugated | |
CRIPT | |
1200020A08Rik; AA589469; CRIPT; CXXC repeat containing interactor of PDZ3 domain; cysteine-rich interactor of PDZ three; Cysteine-rich interactor of PDZ3; cysteine-rich PDZ-binding protein; HSPC139; postsynaptic protein CRIPT; SSMDF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9419 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P021 | |
CRIPT | |
Recombinant Human CRIPT. Recombinant protein control fragment (Product #RP-106730). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction