Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRIPTO Mouse anti-Human, Clone: 2B11, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM2141M

Description
Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.Specifications
CRIPTO | |
Monoclonal | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P13385 | |
TDGF1 | |
KLH conjugated synthetic peptide derived from human Teratocarcinoma-derived growth factor 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B11 | |
Unconjugated | |
TDGF1 | |
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; Cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; TDGF1; TDGF2; teratocarcinoma-derived growth factor; Teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR) | |
Mouse | |
Protein G | |
RUO | |
6997 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction