Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cripto Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517191 Supplier Invitrogen™ Supplier No. PA517191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cripto
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 151 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene TDGF1
Gene Accession No. P51865
Gene Alias CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; epidermal growth factor-like cripto protein CR1; Tdgf1; TDGF2; teratocarcinoma-derived growth factor; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR)
Gene Symbols TDGF1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids near the amino terminus of mouse Cripto.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21667
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.