Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISP-1 Antibody [DyLight 594], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4675M
Description
CRISP-1 Polyclonal antibody specifically detects CRISP-1 in Mouse samples. It is validated for Immunohistochemistry,Western BlotSpecifications
CRISP-1 | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
167 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Western Blot | |
DyLight 594 | |
Acidic epididymal glycoprotein homolog, acidic epididymal glycoprotein-like 1, AEG-related protein, ARPAEG-like protein, CRISP-1AEGL1HSCRISP1D, cysteine-rich secretory protein 1, cysteine-rich secretory protein-1 delta, HSCRISP1G, HUMARP | |
Mouse myeloma cell line NS0-derived recombinant mouse CRISP-1, Gln20-His244, Accession # Q03401 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction