Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRISP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107108
Description
Sequence homology: Cow: 85%; Dog: 85%; Goat: 85%; Human: 100%; Pig: 85%; Rabbit: 79%; Zebrafish: 100%.
May have a role in sperm-egg fusion and maturation.
Specifications
| CRISP1 | |
| Polyclonal | |
| Unconjugated | |
| CRISP1 | |
| Acidic epididymal glycoprotein homolog; acidic epididymal glycoprotein-like 1; AEGL1; AEG-like protein; AEG-related protein; ARP; CRISP1; CRISP-1; cysteine rich secretory protein 1; cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P54107 | |
| CRISP1 | |
| Synthetic peptide directed towards the N terminal region of human CRISP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction