Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRISP1 Polyclonal Antibody

Catalog No. PIPA5122066
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122066 20 μL
PIPA5119066 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122066 Supplier Invitrogen™ Supplier No. PA5122066
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CRISP1 Polyclonal Antibody for Western Blot

May have a role in sperm-egg fusion and maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CRISP1
Gene Accession No. P12020, Q03401
Gene Alias Acidic epididymal glycoprotein homolog; acidic epididymal glycoprotein-like 1; AEGL1; AEG-like protein; AEG-related protein; ARP; CRISP1; CRISP-1; cysteine rich secretory protein 1; cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP
Gene Symbols CRISP1
Host Species Rabbit
Immunogen Recombinant protein Cysteine Rich Secretory Protein 1. The antigen corresponds to amino acid range 20-244 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11571, 654517
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.