Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRISP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597621 Supplier Invitrogen™ Supplier No. PA597621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRISP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The cysteine-rich secretory proteins (CRISP) family is a group of four proteins that are strongly expressed in the male reproductive tract and have been implicated in having roles in male fertility. CRISP2, also known as TPX1, has been implicated in the adhesion between spermatids and Sertoli cells, and with CRISP1, is thought to be involved in sperm-egg fusion. CRISP2 has been shown to regulate the Ca2+ influx through ryanodine receptors (RYR) and may influence the acrosome reaction or sperm motility. CRISP2 has also been shown to bind to the mitogen-activated protein kinase kinase kinase 11 (MAP3K11) and localizes to the developing acrosome, suggesting this CRISP2-MAP3K11 complex may have a role in acrosome development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRISP2
Gene Accession No. P16562
Gene Alias Cancer/testis antigen 36; CRISP2; CRISP-2; Crisp2l; CT36; cysteine rich secretory protein 2; cysteine-rich secretory protein 2; cysteine-rich secretory protein 2-like; GAPDL5; glyceraldehyde-3-phosphate dehydrogenase-like 5; MGC111136; OTTHUMP00000016589; testicular tissue protein Li 43; testis specific gene 1; testis specific protein 1; testis specific protein 1 (probe H4-1 p3-1); testis-specific protein TPX-1; TPX1; Tpx-1; TSP1
Gene Symbols CRISP2
Host Species Rabbit
Immunogen Recombinant Human Cysteine-rich secretory protein 2 protein (22-243AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360445, 7180
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.