Learn More
Invitrogen™ CRISP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109590
Description
Immunogen sequence: KDPAFTALLT TQLQVQREIV NKHNELRKAV SPPASNMLKM EWSREVTTNA QRWANKCTLQ HSDPEDRKTS TRCGENLYMS SDPTSWSSAI QSWYDEILDF VYGVGPKSPN AVVGHYTQLV WYSTYQVGCG IAYCPNQDSL KYYYVCQYCP AGNNMNRKNT PYQQGTPCAG CPDDCDKGLC TNSCQYQDLL SNCDSLKNTA GCEHELLKEK CKATCLCENK IY.
The cysteine-rich secretory proteins (CRISP) family is a group of four proteins that are strongly expressed in the male reproductive tract and have been implicated in having roles in male fertility. CRISP2, also known as TPX1, has been implicated in the adhesion between spermatids and Sertoli cells, and with CRISP1, is thought to be involved in sperm-egg fusion. CRISP2 has been shown to regulate the Ca2+ influx through ryanodine receptors (RYR) and may influence the acrosome reaction or sperm motility. CRISP2 has also been shown to bind to the mitogen-activated protein kinase kinase kinase 11 (MAP3K11) and localizes to the developing acrosome, suggesting this CRISP2-MAP3K11 complex may have a role in acrosome development.
Specifications
CRISP2 | |
Polyclonal | |
Unconjugated | |
CRISP2 | |
Cancer/testis antigen 36; CRISP2; CRISP-2; Crisp2l; CT36; cysteine rich secretory protein 2; cysteine-rich secretory protein 2; cysteine-rich secretory protein 2-like; GAPDL5; glyceraldehyde-3-phosphate dehydrogenase-like 5; MGC111136; OTTHUMP00000016589; testicular tissue protein Li 43; testis specific gene 1; testis specific protein 1; testis specific protein 1 (probe H4-1 p3-1); testis-specific protein TPX-1; TPX1; Tpx-1; TSP1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22024, 360445 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16563 | |
CRISP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-243 of human CRISP2 (NP_003287.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.