Learn More
Invitrogen™ CRISP3 Monoclonal Antibody (295203)

Description
In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant human CRISP-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CRISP3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 295203 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Crisp3 |
| Gene Accession No. | P54108 |
| Gene Alias | 32 kDa epididymal protein; Acidic epididymal glycoprotein; Acidic epididymal glycoprotein 2; acidic epididymal glycoprotein D/E; Aeg; Aeg2; Aeg-2; Crisp1; Crisp-1; Crisp3; CRISP-3; CRISP-3MGC126588; CRS3; cysteine rich secretory protein 3; cysteine-rich secretory protein 1; Cysteine-rich secretory protein 3; cysteine-rich secretory protein-3; dJ442L6.3; epididymal glycoprotein; protein D; protein E; protein IV; SCP; SCP 2; SGP28; Sialoprotein; specific granule protein (28 kDa); specific granule protein of 28 kDa; Sperm-coating glycoprotein; sperm-coating glycoprotein 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.