Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRISP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584197 Supplier Invitrogen™ Supplier No. PA584197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCPDNCDDGL CTNGCKYEDL YSNCKSLKLT LTCKHQLVRD SCKASCNCSN S Highest antigen sequence indentity to the following orthologs: Mouse - 57%, Rat - 57%.

Cysteine-rich secretory protein 3 (CRISP-3; also known as SGP28) is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Crisp3
Gene Accession No. P54108
Gene Alias 32 kDa epididymal protein; Acidic epididymal glycoprotein; Acidic epididymal glycoprotein 2; acidic epididymal glycoprotein D/E; Aeg; Aeg2; Aeg-2; Crisp1; Crisp-1; Crisp3; CRISP-3; CRISP-3MGC126588; CRS3; cysteine rich secretory protein 3; cysteine-rich secretory protein 1; Cysteine-rich secretory protein 3; cysteine-rich secretory protein-3; dJ442L6.3; epididymal glycoprotein; protein D; protein E; protein IV; SCP; SCP 2; SGP28; Sialoprotein; specific granule protein (28 kDa); specific granule protein of 28 kDa; Sperm-coating glycoprotein; sperm-coating glycoprotein 2
Gene Symbols Crisp3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRISP3. Recombinant protein control fragment (Product #RP-100095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.