Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987308
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7308 Supplier Bioss Supplier No. BS6521R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cysteine-rich secretory protein 3 (CRISP-3; also known as SGP28) is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.

Specifications

Antigen CRISP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Crisp3
Gene Accession No. P54108
Gene Alias Acidic epididymal glycoprotein 2; Aeg2; Aeg-2; Crisp3; CRISP-3; CRISP-3MGC126588; CRS3; cysteine rich secretory protein 3; Cysteine-rich secretory protein 3; cysteine-rich secretory protein-3; dJ442L6.3; SCP 2; SGP28; specific granule protein (28 kDa); Specific granule protein of 28 kDa; Sperm-coating glycoprotein 2
Gene Symbols Crisp3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRISP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10321
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.