Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRISPLD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555162
Description
Immunogen sequence: ACPPSFGGGC RENLCYKEGS DRYYPPREEE TNEIERQQSQ VHDTHVRTRS DDSSRNEVIS AQQMSQIVSC EVRLRDQCKG TTCNRYECPA GC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 92%.
Specifications
| CRISPLD1 | |
| Polyclonal | |
| Unconjugated | |
| CRISPLD1 | |
| CocoaCrisp; CRISP10; CRISP-10; CRISPLD1; cysteine rich secretory protein LCCL domain containing 1; cysteine-rich secretory protein 10; cysteine-rich secretory protein LCCL domain containing 1; cysteine-rich secretory protein LCCL domain-containing 1; LCCL domain containing cysteine-rich secretory protein 1; LCCL domain-containing cysteine-rich secretory protein 1; LCRISP1; RGD1564813; Trypsin inhibitor Hl; UNQ342/PRO541 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H336 | |
| CRISPLD1 | |
| Recombinant protein corresponding to Human CRISPLD1. Recombinant protein control fragment (Product #RP-93774). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction