Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPLD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169620
Description
CRISPLD2 Polyclonal specifically detects CRISPLD2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CRISPLD2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CRISP11, CRISP-11, Cysteine-rich secretory protein 11, cysteine-rich secretory protein LCCL domain containing 2, cysteine-rich secretory protein LCCL domain-containing 2, DKFZP434B044, LCCL domain containing cysteine-rich secretory protein 2, LCCL domain-containing cysteine-rich secretory protein 2, LCRISP2, MGC74865 | |
Rabbit | |
56 kDa | |
100 μL | |
Primary | |
Mouse: 86%. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunoprecipitation | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunoprecipitation | |
Q9H0B8 | |
CRISPLD2 | |
Synthetic peptides corresponding to CRISPLD2(cysteine-rich secretory protein LCCL domain containing 2) The peptide sequence was selected from the N terminal of CRISPLD2. Peptide sequence MSCVLGGVIPLGLLFLVCGSQGYLLPNVTLLEELLSKYQHNESHSRVRRA. | |
Affinity purified | |
RUO | |
83716 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction