Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPLD2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297242PCP
Description
CRISPLD2 Polyclonal antibody specifically detects CRISPLD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CRISPLD2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CRISP11, CRISP-11, Cysteine-rich secretory protein 11, cysteine-rich secretory protein LCCL domain containing 2, cysteine-rich secretory protein LCCL domain-containing 2, DKFZP434B044, LCCL domain containing cysteine-rich secretory protein 2, LCCL domain-containing cysteine-rich secretory protein 2, LCRISP2, MGC74865 | |
Produced in rabbits immunized with E. coli-derived Human CRISPLD2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
83716 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction