Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRISPLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589067 Supplier Invitrogen™ Supplier No. PA589067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPYPSECNPW CPERCSGPMC THYTQIVWAT TNKIGCAVNT CRKMTVWGEV WENAVYFVCN YSPKGNWIGE APYKNGRPCS ECPPSYGGSC RNNLCYREET YTPKPETDEM NEVETAPIPE ENHVWLQPRV MRPTKPKKTS AVNYMTQVVR CDTKMKDRCK GSTCNRYQCP AGCLNHKAKI FGTLFYESSS SICRAAIHYG ILDDKGGLVD ITRNGKVPFF VKSERHGVQS LSKYKPSSSF MVSKVKVQDL DCYTTVAQLC PFEKPATHCP RIHCPAHCKD EPSYWAPVFG T; Positive Samples: Mouse kidney, Mouse pancreas, Rat pancreas.

CRISPLD2 promotes matrix assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISPLD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CRISPLD2
Gene Accession No. Q8BZQ2
Gene Alias 1810049K24Rik; coffeecrisp; CRISP11; CRISP-11; CRISPLD2; cysteine rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein 11; cysteine-rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein LCCL domain-containing 2; late gestation lung protein 1; LCCL domain containing cysteine-rich secretory protein 2; LCCL domain-containing cysteine-rich secretory protein 2; Lcrisp2; Lgl1; testis secretory sperm-binding protein Li 207a; trypsin inhibitor; UNQ2914/PRO1156/PRO9783
Gene Symbols CRISPLD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 140-430 of human CRISPLD2 (NP_1136641).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171547, 78892
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.