Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRISPLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110992 Supplier Invitrogen™ Supplier No. PA5110992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRNGKVPFFV KSERHGVQSL SKYKPSSSFM VSKVKVQDLD CYTTVAQLCP FEKPATHCPR IHCPAHCKDE PSYWAPVFGT NIYADTSSIC KTAVHAGVIS NESGGDVDVM PVDK Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 78%.

CRISPLD2 promotes matrix assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISPLD2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRISPLD2
Gene Accession No. Q9H0B8
Gene Alias 1810049K24Rik; coffeecrisp; CRISP11; CRISP-11; CRISPLD2; cysteine rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein 11; cysteine-rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein LCCL domain-containing 2; late gestation lung protein 1; LCCL domain containing cysteine-rich secretory protein 2; LCCL domain-containing cysteine-rich secretory protein 2; Lcrisp2; Lgl1; testis secretory sperm-binding protein Li 207a; trypsin inhibitor; UNQ2914/PRO1156/PRO9783
Gene Symbols CRISPLD2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CRISPLD2. Recombinant protein control fragment (Product #RP-108330).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.