Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody (6H4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281124APC
Description
CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
CRISPR-Cas9 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |
Western Blot, Immunocytochemistry, Immunoprecipitation | |
6H4 | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
This CRISPR-Cas9 antibody (6H4) was raised against partial recombinant protein made to an N terminal portion of the Staphylococcus aureus CRISPR/Cas9 protein | |
0.1 mL | |
Gene Editing, Gene Knockouts, Gene Therapy | |
901176 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction