Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody (6H4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281124B
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CRISPR-Cas9 | |
| Monoclonal | |
| Biotin | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| Partial recombinant protein made to an N terminal portion of the Staphylococcus aureus CRISPR/Cas9 protein | |
| 0.1 mL | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| 901176 | |
| Store at 4°C in the dark. |
| Western Blot, Immunofluorescence | |
| 6H4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction