Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Antibody (7A9-3A3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP236440AF532

Catalog No. NP236440A32


Only null left
Add to Cart

Description

Description

CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Mouse, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated.
Specifications

Specifications

CRISPR-Cas9
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
Primary
This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus is specific toCas9 protein from Streptococcus pyogene serotype M1.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoblot, Immunoassay
7A9-3A3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2].
Gene Editing, Gene Knockouts, Gene Therapy
901176
Mouse, Bacteria
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.