Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4964513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236440SS 0.025 mL
NBP236440 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236440SS Supplier Novus Biologicals Supplier No. NBP236440SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, Immunoassay.

Specifications

Antigen CRISPR-Cas9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunoassay
Classification Monoclonal
Clone 7A9-3A3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Frozen reported in scientific literature (PMID 28153089), Immunoblotting reported in scientific literature (PMID 31959836), Immunohistochemistry (Whole-Mount), Knockout Validated reported in scientific literature (PMID 31959836)
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease, SpyCas9
Host Species Mouse
Immunogen This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2].
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Test Specificity Cas9 protein from Streptococcus pyogene serotype M1.
Target Species Bacteria, Chicken, Fungi, Fungi
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.