Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | CRISPR-Cas9 |
---|---|
Concentration | 1.0mg/mL |
Dilution | Western Blot 2μg/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP266379SS
![]() |
Novus Biologicals
NBP266379SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP266379
![]() |
Novus Biologicals
NBP266379 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.Specifications
CRISPR-Cas9 | |
Western Blot 2μg/ml | |
Polyclonal | |
Rabbit | |
Bacteria | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
IgG | |
Affinity Purified | |
Cas9 protein from Streptococcus pyogene serotype M1. |
1.0mg/mL | |
Western Blot | |
Unconjugated | |
Gene Editing, Gene Knockouts, Gene Therapy | |
PBS with 0.02% Sodium Azide | |
Full length S. pyogenes Cas9 protein [UniProt Q99ZW2] | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title