Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody (JM11-55), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267000
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in S. pyogenes, Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CRISPR-Cas9 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
Recombinant protein within Streptococcus pyogenes CRISPR-Cas9 aa 730-990/ 1368. (SwissProt: Q99ZW2 StreptococcusPyogenes) | |
Protein A purified | |
Gene Editing, Gene Knockouts, Gene Therapy | |
S. pyogenes, Bacteria | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JM11-55 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
Rabbit | |
158 kDa | |
100 μL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction