Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody (SR1104), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
| Antigen | CRISPR-Cas9 |
|---|---|
| Clone | SR1104 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
Description
CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in S. aureus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| CRISPR-Cas9 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| S. aureus | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
| A synthesized peptide derived from CRISPR-Cas9 (Uniprot #: J7RUA5) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1104 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 901176 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title