Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cpf1 Antibody (3D3-F7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP278820AF405
Description
CRISPR-Cpf1 Monoclonal specifically detects CRISPR-Cpf1 in Bacterial samples. It is validated for Western Blot.Specifications
CRISPR-Cpf1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
This antibody is specific for the Acidaminococcus sp. CRISPR-Cpf1 | |
Store at 4°C in the dark. |
Western Blot | |
3D3-F7 | |
Western Blot | |
Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
Partial recombinant protein made to a C-terminal portion of the Acidaminococcus sp. CRISPR-Cpf1 protein | |
0.1 mL | |
Primary | |
Bacteria | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction