Learn More
AlphaProTech Critical Cover™ CoolOne™ Sensitive Skin Face Masks
BIOPHARMA
Catalog No. p-7237199 Shop All AlphaProTech Products
Click to view available options
Includes:
Anti-fog earloops
Soft knit earloops
This item is not returnable. View return policy
This item is not returnable. View return policy
Fluid resistant face masks are free of pigments and dyes and are available in two earloop styles. Patented Magic Arch™ support strip holds the mask away from the nose and mouth to create a larger breathing chamber.
  • Fluid resistant face masks are free of pigments and dyes
  • Patented Magic Arch™ support strip holds mask away from the nose and mouth to create a larger breathing chamber
  • Two styles to choose from:
    • Soft knit earloops
    • Anti-fog ear loops for use in high humidity environments or with protective eyewear
  • ASTM F2100 Compliant Level 1 mask
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color White
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Includes Anti-fog earloops
Model WH 6155
Packaging Quantity 500/Cs.
Pore Size 0.1 μm
Resistance Fluid
Series CoolOne™
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.